Learn More
Chemglass Life Sciences Chemglass Pipette Tips, Clear, Non-Sterile, 1250 μL, Bulk Nest (CLS-3499E-125)
SDP
Supplier:  Chemglass Life Sciences CLS3499E125
Encompass_Preferred
Chemglass 1250 μL clear pipette tips are large-volume, ultra-clear polypropylene tips supplied non-sterile in nested bulk packaging for high-throughput laboratory pipetting. They are designed for minimal sample adhesion and broad compatibility with most single and multichannel pipettors.
  • Ultra-clear polypropylene construction.
  • Non-sterile and certified RNase-, DNase-, human DNA-, and pyrogen-free.
  • Low-retention, fine-point design for reduced sample adhesion.
  • Bulk nested packaging for high-volume workflows.
  • Universal-fit for most single and multichannel pipettors.
  • Pack size options: 1,000 per pack, 10,000 per case.
Catalog No. 50-273-3209