Learn More
Pacer Global LLC 125mL Erlenmeyer Flask, Vent Cap, Plain Bottom, PETG, Sterile, 12 per Case
SDP
Supplier:  Pacer Global LLC MLSEFVC125/DEL
Encompass_Preferred
125mL PETG Erlenmeyer flasks are designed for mammalian suspension culture applications as well as bacterial & yeast culture. Packaged Sterile in individual bags for easy storage and handling. They have a plain bottom with an HDPE vented 0.22µm hydrophobic membrane which allows for improved gas exchange without contamination cap. Autoclavable. Individually Packed. 12/case.
Catalog No. NC1862170