Learn More
Integra Biosciences Corp 50/125 µl VIAFLO/VOYAGER pipettes, 5 racks of 384 GripTips
SDP
Supplier:  Integra Biosciences Corp 4425
Encompass
GripTips for use with VIAFLO Pipettes. Sterile filter; 50ul/125ul; 5 Racks of 384 tips. GripTips effortlessly snap onto the tip fittings and are easily ejected. GripTips feature a high lateral resistance once attached to the VIAFLO II pipettes.
Catalog No. NC0043753
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product