Learn More
CSAFE LLC CSafe Global 12L Refrigerated Courier Solution for 2°C to 8°C. Maintains 2°C to 8°C for 12 hours.
Supplier:  CSAFE LLC PX12LU28
Encompass
CSafe Courier solutions are designed for physicians, pharmacists, researchers, pharmaceutical representatives, and other healthcare professionals making last-mile delivery of temperature-sensitive life science products. Maintains temperature for 12 hours.Product Storage Temperature: 2°C to 8°C (36°F to 46°F) | Payload Volume: 12.0L (758.6in³) | Internal Payload Dimensions mm (inches): 216 x 216 x 267 (8.5 x 8.5 x 10.5).
Catalog No. NC1726160