Learn More
Cole-Parmer Fowler 52-264-012 52-264012: 12" /300MM Pre-Certified Flexible Rule
Supplier:  Cole-Parmer 0991916
Encompass_Preferred
Certified rulers are made of hardened and tempered steel and feature easy to read graduations on a non-glare satin chrome finish. Each Fowler Ruler is Pre-Certified for ISO/QS9000 requirements and includes a certificate of calibration.
Catalog No. NC9655854