Learn More
HARVARD BIOSCIENCE INC 12MM DIAM NO1 COVERGLASS
Supplier:  HARVARD BIOSCIENCE INC 640702
Encompass_Preferred
Total order minimum requirement of $500. If only item on order, must order QTY of 11.Standard Dimension Coverslips: #1 Thickness (0.15 0.02 mm)
Catalog No. NC9702891