Learn More
Enzo Life Sciences 12(S)-Hydroxyeicosa-5Z,8Z,10E,14Z-tetraenoic acid (HETE) (50µg). CAS: 54397-83-0
SDP
Supplier:  Enzo Life Sciences BMLH0120050
Encompass_Preferred
Biosynthesis by platelet 12-lipoxygenase. Chemotactic and chemokinetic for PMNL2 and for vascular smooth muscle cells. Antiproliferative effect on smooth muscle cells. Second messenger in angiotensin induced aldosterone production. Induces endothelial cell retraction and tumor cell adhesion. Alternative name: 12(S)-HETE. Formulation: Liquid. Solution in ethanol. Purity: ≥98% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-1018