Learn More
Altenloh Brinck & Co US INC. DBA: SABEU 12 well insert 1.0um PET 4Plate (cellQART)
Supplier:  Altenloh Brinck & Co US INC. 9311014
Encompass_Preferred
cellQART Cell Culture Inserts combined with our especially designed Well Plates are intended to be used for cell culture and in vitro tissue culture research applications. The Well Plate is supplied in 12-Well format, it is sterile and fully pre-loaded with cellQART Cell Culture Inserts. These cellQART Cell Culture Inserts come with PET membranes in clear optics. The Inserts are tissue culture treated and sterile. They are ideal for in-situ phase contrast microscopy.
Catalog No. 50-267-3503