Learn More
Andwin Scientific 12X12 CERAMIC HOTPLATE 115V EA
SDP
Supplier:  Andwin Scientific 480114
Encompass_Preferred
The 115V hot plate features a 12x12 corrosion-resistant ceramic top ideal for high temperature applications housed in an aluminum frame for cooler edges. It also boast an epoxy-coated cast-aluminum body for maximum durability and stability. The temperature range runs from 49 to 450C. The unit includes a 6ft cord with plug.
Catalog No. 50-130-5913