Learn More
US Plastic Corporation 0.118" (3.0mm) x 12" x 24" Blue Fluorescent Acrylic Sheet
SDP
Supplier:  US Plastic Corporation 86201
Encompass
Great for many different projects and are easy to cut, drill, and polish to whatever shapes you may need. They are perfect for thousands of jobs. These colors are very light and transparent-not bright, brilliant colors normally associated with fluorescent colors. Not UV resistant. Indoor use only.
Catalog No. NC9915003