Learn More
Cayman Chemical 13 14dIhydro PROSGlnDN F 10mg
SDP
Supplier:  Cayman Chemical 1561010 MG
Encompass_Preferred
13,14-dihydro PGF1α is a potential metabolite of PGF1α.{2346} Its biological activity has not been reported. The corresponding compound in the E1 series (13,14-dihydro PGE1) retains biological activity similar to PGE1.{4168}
Catalog No. 50-296-9689