Learn More
Cayman Chemical 13 14dIhydro15 R Shydroxy 50UG
SDP
Supplier:  Cayman Chemical 900040650 UG
Encompass_Preferred
An internal standard for quantification of 13,14-dihydro-15 (R,S)-hydroxy-16,16-difluoro-Prostaglandin E1, a metabolite of a bicyclic PGE1 derivative used in the treatment of chronic constipation, by GC- or LC-mass spectrometry (MS)
Catalog No. 50-289-5611