Learn More
Cayman Chemical 13 14epoxy FlupROSnl Isop 10mg
SDP
Supplier:  Cayman Chemical 1367910 MG
Encompass_Preferred
An impurity generated in the production of travoprost; a 4:96 diastereomeric mixture of epoxides
Catalog No. 50-295-3416