Learn More
Medchemexpress LLC 13-cis-4-oxoretinoic acid | 71748-58-8 | MFCD00870452 | >98.0% | 314.42 g/mol | C20H26O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY107494100MG
Encompass_Preferred
13-cis-4-oxoretinoic acid is an oxidized metabolite of vitamin A used as an analytical reference standard and research reagent in biochemical and pharmacological studies. CAS 71748-58-8; formula C20H26O3; molecular weight 314.42 g/mol. High-purity material is used for quantitative and qualitative assays.
  • Used as a reference standard for analytical and methodological research.
  • Relevant to vitamin A metabolism studies and metabolite profiling.
  • Provided as a small-molecule solid suitable for laboratory handling.
  • High purity supports quantitative and qualitative assays.
  • Compatible with chromatographic and mass spectrometric methods.
Catalog No. 50-002-41432