Learn More
TARGETMOL CHEMICALS INC 13-Methylberberine chloride
SDP
Supplier:  TARGETMOL CHEMICALS INC TN11895MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 13-Methylberberine chloride shows anti-adipogenic effect on 3T3-L1 adipocytes, it has potential as an anti-obesity drug Purity 99.85%
Catalog No. 50-287-7171