Learn More
JANT PHARMACAL CORPORATION ACCUTEST Drug Test Cup 13-Panel CLIA-Waived: MTD/OXY/PCP/BAR/BUP/BZO/MET/THC/TCA/COC/MOR(300)/AMP/XTC 25 tests/box
SDP
Supplier:  JANT PHARMACAL CORPORATION DS823A
Encompass_Preferred
The ACCUTEST 13 panel multi drug screen is an easy, fast, qualitative, visually read competitive binding immunoassay method for screening without the need of instrumentation. The cup is FDA (510) K Cleared and CLIA Waived. The 13 panel CLIA Waived drug test cup screens for the presence of Methadone, Oxycodone, Phencyclidine, Barbiturate, Buprenorphine, Benzodiazepine, Methamphetamine, Marijuana, Tricyclic, Cocaine, Opiates, Amphetamine and Ecstasy in human urine, which is qualitativly analzyed to indicate a positive or negative test result.
Catalog No. NC3457092