Learn More
Cayman Chemical 13 S-HODE-d4 50ug
SDP
Supplier:  Cayman Chemical 33861050 UG
Encompass_Preferred
An internal standard for the quantification of 13-HODE by GC- or LC-MS
Catalog No. 50-294-8136