Learn More
Cayman Chemical 13 S-HpOTrE 100ug
SDP
Supplier:  Cayman Chemical 45220100 UG
Encompass_Preferred
A monohydroperoxy PUFA produced by the action of 15-LO on α-linolenic acid
Catalog No. 50-297-8314