Learn More
Sino Biological Recombinant Human P-Selectin/CD62P Protein (hFc Tag)
Supplier:  Sino Biological 13025H02H100
Encompass_Preferred
A DNA sequence encoding the human SELP (AAN06828.1) extracellular domain (Met 1-Ala 771) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8172