Learn More
Sino Biological Recombinant Human Tissue Factor Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 13133H02H20
Encompass_Preferred
A DNA sequence encoding the human F3 (P13726) extracellular domain (Met 1-Glu 251) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8307