Learn More
Sino Biological Recombinant Human TROY/TNFRSF19 Protein (His Tag)
Supplier:  Sino Biological 13387H08H100
Encompass_Preferred
A DNA sequence encoding the human TNFRSF19 (Q9NS68-2) (Met1-Leu170) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8548