Learn More
WELCH MATERIALS INC DoprahTM 13-425 Black PP Open Top Screw Cap with Red PTFE/White Silicone Septa 1mm Thick. 100pcs/pk, 10pk/cs
Supplier:  WELCH MATERIALS INC DC117910
Encompass_Preferred
13-425 Black PP Open Top Screw Cap with Red PTFE/White Silicone Septa 1mm Thick. 100pcs/pk, 10pk/cs
Catalog No. 50-209-3145