Learn More
Sino Biological Recombinant Human IGSF8 Protein (His Tag)
Supplier:  Sino Biological 13435H08H50
Encompass_Preferred
A DNA sequence encoding the human IGSF8 (Q969P0-1) (Met1-Thr579) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8573