Learn More
Sino Biological Recombinant Human LYG1 Protein (His Tag)
Supplier:  Sino Biological 13457H08B100
Encompass_Preferred
A DNA sequence encoding the human LYG1 (Met 1-Phe194) (Q8N1E2) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-8588