Learn More
Electron Microscopy Sciences Flim Sleeve 135 mm/20 exp with Frosted Open Flap
SDP
Supplier:  Electron Microscopy Sciences 73933
Encompass_Preferred
Polyguard Polypropylene film sleeves are the finest archival quality material, and are guaranteed to give you ultimate protection for your most important film negatives. Polyguard is 100% acid free, as well as being free from optical distortion. It is resistant to water, grease, oil, alkalies, and organic solvents. Polyguard is durable, yet pliable and gentle to the film and will not become brittle, crack, or abrade the film.
Type: Frosted Open Flap
Catalog No. 50-280-15