Learn More
Sino Biological Recombinant Human GITR/TNFRSF18/AITR Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 13643H08H100
Encompass_Preferred
A DNA sequence encoding the human TNFRSF18 (NP_683700.1) (Met1-Glu161) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8726