Learn More
Sino Biological Recombinant Human EPDR1 Protein (His Tag)
Supplier:  Sino Biological 13665H08H50
Encompass_Preferred
A DNA sequence encoding the human EPDR1 (Q9UM22-1) (Met1-Trp224) with a C-terminal polyhistidine tag was expressed.
Catalog No. 50-161-8762