Learn More
Medchemexpress LLC HY-112514 5mg Medchemexpress, 1,3,6,8-Tetrahydroxynaphthalene CAS:18512-30-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1125145MG
Encompass_Preferred
Medchemexpress, HY-112514 5mg 1,3,6,8-Tetrahydroxynaphthalene CAS:18512-30-6 1,3,6,8-Tetrahydroxynaphthalene (T4HN) is an indispensable precursor to DHN (1,8-Dihydroxynaphthalene) melanin and is an unique symmetrical compound of polyketide origin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0818