Learn More
eMolecules​ 136866-27-8 | 3,5-DICHLOROPYRAZINE-2-CARBALDEHYDE | AstaTech | MFCD09864908 | 176.980 | C5H2Cl2N2O | 95.000 | Clc1cnc(C=O)c(Cl)n1 | 0.25g | 449769410
Supplier:  eMolecules​ 449769410
Encompass_Preferred
3,5-DICHLOROPYRAZINE-2-CARBALDEHYDE | AstaTech | 136866-27-8 | MFCD09864908 | 176.980 | C5H2Cl2N2O | 95.000 | Clc1cnc(C=O)c(Cl)n1 | 0.25g | 449769410
Catalog No. 50-004-92825