Learn More
eMolecules​ 13734-36-6 | Boc-Sar-OH | AA Blocks LLC | MFCD00037795 | 189.211 | C8H15NO4 | 0.000 | CN(CC(O)=O)C(=O)OC(C)(C)C | 10g | 437040691
Supplier:  eMolecules​ 437040691
Encompass_Preferred
Boc-Sar-OH | AA Blocks LLC | 13734-36-6 | MFCD00037795 | 189.211 | C8H15NO4 | 0.000 | CN(CC(O)=O)C(=O)OC(C)(C)C | 10g | 437040691
Catalog No. 50-004-92892