Learn More
Sino Biological Recombinant Human LMAN2L Protein (hFc Tag)
Supplier:  Sino Biological 13822H02H100
Encompass_Preferred
A DNA sequence encoding the human LMAN2L (NP_110432.1) (Met1-Ala304) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8853