Learn More
Sino Biological Recombinant Human Epiphycan Protein (His Tag)
Supplier:  Sino Biological 13855H08H50
Encompass_Preferred
A DNA sequence encoding the human EPYC (AAH30958.1) (Met1-Val322) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8870