Learn More
eMolecules​ 139139-86-9 | (R)-H8-BINAP | Ambeed | MFCD01630795 | 630.752 | C44H40P2 | 98.000 | C1CCc2c(C1)ccc(P(c1ccccc1)c1ccccc1)c2-c1c2CCCCc2ccc1P(c1ccccc1)c1ccccc1 | 5g | 686016175
Supplier:  eMolecules​ 686016175
Encompass_Preferred
(R)-H8-BINAP | Ambeed | 139139-86-9 | MFCD01630795 | 630.752 | C44H40P2 | 98.000 | C1CCc2c(C1)ccc(P(c1ccccc1)c1ccccc1)c2-c1c2CCCCc2ccc1P(c1ccccc1)c1ccccc1 | 5g | 686016175
Catalog No. 50-004-90835