Learn More
Sino Biological Recombinant Human Factor H-related 2/CFHR2 Protein (His Tag)
Supplier:  Sino Biological 13920H08H50
Encompass_Preferred
A DNA sequence encoding the human CFHR2 (P36980-1) (Met1-Lys270) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8928