Learn More
Cayman Chemical 13acetyl9DIhydrobaccatIn I 1mg
SDP
Supplier:  Cayman Chemical 297011 MG
Encompass_Preferred
A taxane with anticancer activity; inhibits proliferation of P388 leukemia cells (IC50 = 20 μg/ml)
Catalog No. 50-298-8930