Learn More
Cayman Chemical 13C6-4-NItrophenylhydrazIn 1mg
SDP
Supplier:  Cayman Chemical 404791 MG
Encompass_Preferred
An internal standard for the quantification of 4-nitrophenylhydrazine by GC- or LC-MS; a byproduct in the synthesis of 13C6-3-nitrophenylhydrazine
Catalog No. 50-294-4671