Learn More
Cayman Chemical 13C6-6-PPD-quInon 5mg
SDP
Supplier:  Cayman Chemical 406065 MG
Encompass_Preferred
An internal standard for the quantification of 6-PPD-quinone by GC- or LC-MS
Catalog No. 50-294-5452