Learn More
Cayman Chemical 13C6 GlucosylsphngosInd18 1mg
SDP
Supplier:  Cayman Chemical 232121 MG
Encompass_Preferred
An internal standard for the quantification of 1-β-D-glucosylsphingosine by GC- or LC-MS
Catalog No. 50-299-3790