Learn More
Enzo Life Sciences 13(S)-Hydroperoxyoctadeca-9Z,11E-dienoic acid (50µg). CAS: 33964-75-9
SDP
Supplier:  Enzo Life Sciences BMLHP0130050
Encompass_Preferred
Suppresses activation of platelets. Induces endothelial injury. Constricts isolated coronary arteries. Inhibits prostacyclin synthase. Alternative name: 13(S)-HPODE. Formulation: Oil dissolved in ethanol. Purity: ≥98%. Long Term Storage: -80°C.
Catalog No. 50-201-1033