Learn More
Sarstedt Inc Screw cap tube, 5 ml, (LxO): 75 x 13 mm, round base, PP, 1000/pk
Supplier:  Sarstedt Inc 60.504.031/DUP
Encompass_Preferred
Screw cap tube, working volume: 5 ml, (LxO): 75 x 13 mm, round base, transparent, material: PP, without cap.
Catalog No. NC1866722
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.