Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 14,15-EE-5(Z)-E | 519038-92-7 | >98.0% | 25 UG
SDP

Supplier:  Medchemexpress LLC HY12095425UG

Encompass_Preferred

14,15-EE-5(Z)-E is a structural analogue of 14,15-epoxide dicartrienoic acid (14,15-EET) that antagonizes EET-induced relaxation of vascular smooth muscle. It is used as a biochemical probe in cardiovascular research to study EET-mediated signaling.

  • Structural analogue of 14,15-epoxide dicartrienoic acid (14,15-EET).
  • Antagonizes EET-induced relaxation of vascular smooth muscle.
  • Molecular formula: C20H36O3; molecular weight: 324.50.
  • CAS number: 519038-92-7.
  • Purity typically ≥98% (supplier reported).
  • Storage: solution, -20°C, up to 2 years.
  • Common use: probe in cardiovascular and vascular physiology research.

Catalog No. 50-002-34041


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.