Learn More
Cayman Chemical 14 15-EET methyl ester 50UG
SDP
Supplier:  Cayman Chemical 5065050 UG
Encompass_Preferred
An esterified version of (±)14(15)-EET; more stable for long-term storage and can be readily hydrolyzed to the free acid just before use; equipotent to (±)14(15)-EET in inducing relaxation of isolated bovine coronary arteries precontracted by U-46619
Catalog No. 50-289-6428