Learn More
Cayman Chemical 14 15EETd11 methyl este 100ug
SDP
Supplier:  Cayman Chemical 29383100 UG
Encompass_Preferred
An internal standard for the quantification of 14(15)-EET methyl ester by GC- or LC-MS
Catalog No. 50-298-8270