Learn More
Cayman Chemical 14 15EETSI Fatty Acids and Der
SDP
Supplier:  Cayman Chemical 1000928625 UG
Encompass_Preferred
A metabolically stable methyl sulfonamide analog of 14(15)-EET; equipotent to 14(15)-EET in vascular agonist activity
Catalog No. 50-289-0959