Learn More
Cayman Chemical 14 15EpEDE Fatty Acids and Der
SDP
Supplier:  Cayman Chemical 1000752725 UG
Encompass_Preferred
An EpEDE acid formed from DGLA, either by a strong oxidizing agent or by cytochrome P450 epoxygenases; biological properties are poorly understood
Catalog No. 50-289-0818