Learn More
Ace Glass, Inc. 14/35 PTFE Stopper with Polypropylene Extraction Nut, Solid
SDP
Supplier:  Ace Glass, Inc. 1263610
Encompass
14/35 standard taper PTFE stopper with polypropylene extraction nut for easy removal from outer joints.
Catalog No. 50-977-606