Learn More
Chemglass Life Sciences Replacement Viton O-Rings for 14.4/44 PTFE Plug
SDP
Supplier:  Chemglass Life Sciences CG45710
Encompass_Preferred
Replacement Viton O-Rings for 14.4/44 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.
Catalog No. CG45710