Learn More
Medchemexpress LLC HY-121172 5mg Medchemexpress, 1,4-DPCA CAS:331830-20-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1211725MG
Encompass_Preferred
Medchemexpress, HY-121172 5mg 1,4-DPCA CAS:331830-20-7 1,4-DPCA, a potent prolyl-4-hydroxylase inhibitor, is a collagen hydroxylation inhibitor in human foreskin fibroblasts with an IC50 of 2.4 µM. 1,4-DPCA inhibits prolyl-4-hydroxylases α isoforms stabilizes HIF-1α protein. 1,4-DPCA also inhibits factor inhibiting HIF (FIH) with an IC50 of 60 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0963