Learn More
Cayman Chemical 14-nhydrodIgItoxIgenIn 5mg
SDP
Supplier:  Cayman Chemical 304395 MG
Encompass_Preferred
A cardenolide; reduces the activity of guinea pig heart Na+/K+-ATPase by 15% at 10 µM
Catalog No. 50-299-1668