Learn More
Cliawaived Inc CLIAwaived, Inc. 14 Panel - IDTC II
SDP
Supplier:  Cliawaived Inc CLIAIDTC14BUPA
Encompass_Preferred
(25 Cups)CLIAwaived, Inc. - INSTANT DRUG TEST CUP (IDTC II) with Buprenorphine. This 14-panel, no-leak drug test cup includes BUP and adulterants. This cup tests for AMP500, BAR300, BUP10, BZO300, COC150, EDDP300, MDMA500, MET500, MTD300, OPI300, OXY100, PCP25, TCA1000, THC50 + (OX/pH/SG), (GL/NI/CR).Results in 5 minutes. FDA-cleared and CLIA-waived.
Catalog No. NC1792992